Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SMCO3
Entrez Gene ID
618483
Gene Name
single-pass membrane protein with coiled-coil domains 3
Gene Aliases
C5H12orf69
Chromosome Location
Chr.5: 95082187 - 95100477 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001206296.1 NP_001193225.1 1 - 2 27 103

View Full Product Details