Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LVRN
Entrez Gene ID
525695
Gene Name
laeverin
Gene Aliases
AQPEP
Chromosome Location
Chr.10: 4850650 - 4929715 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001206325.2 NP_001193254.1 12 - 13 2035 83

View Full Product Details