Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PILRA
Entrez Gene ID
100137734
Gene Name
paired immunoglobin like type 2 receptor alpha
Gene Aliases
-
Chromosome Location
Chr.25: 36137410 - 36154870 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001206350.2 NP_001193279.1 6 - 7 869 97
GenBank mRNA BC149130.1 - 6 - 7 869 97

View Full Product Details