Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ARC
Entrez Gene ID
519403
Gene Name
activity regulated cytoskeleton associated protein
Gene Aliases
-
Chromosome Location
Chr.14: 1741147 - 1744556 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001206407.1 NP_001193336.1 1 - 2 1924 113

View Full Product Details