Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LANCL3
Entrez Gene ID
519239
Gene Name
LanC like 3
Gene Aliases
-
Chromosome Location
Chr.X: 105734791 - 105847786 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001206408.1 NP_001193337.1 4 - 5 1426 79

View Full Product Details