Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
WDR25
Entrez Gene ID
527651
Gene Name
WD repeat domain 25
Gene Aliases
-
Chromosome Location
Chr.21: 65267504 - 65416854 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001206428.1 NP_001193357.1 5 - 6 1317 90
GenBank mRNA BC149548.1 - 5 - 6 1318 90

View Full Product Details