Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PSTK
Entrez Gene ID
511744
Gene Name
phosphoseryl-tRNA kinase
Gene Aliases
-
Chromosome Location
Chr.26: 42767546 - 42780840 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001206440.1 NP_001193369.1 3 - 4 661 98
GenBank mRNA BC151821.1 - 3 - 4 662 98
CX954191.1 - 3 - 4 442 98

View Full Product Details