Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
REN
Entrez Gene ID
280909
Gene Name
renin
Gene Aliases
-
Chromosome Location
Chr.16: 1936108 - 1956402 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001206509.1 NP_001193438.1 1 - 2 129 75
GenBank mRNA EV658200.1 - 1 - 2 131 75

View Full Product Details