Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CD2AP
Entrez Gene ID
533188
Gene Name
CD2 associated protein
Gene Aliases
-
Chromosome Location
Chr.23: 20602675 - 20689497 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001206539.1 NP_001193468.1 9 - 10 1075 109
GenBank mRNA BC102654.1 - 9 - 10 1075 109
BF041363.1 - 1 - 2 101 109

View Full Product Details