Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC16A6
Entrez Gene ID
529171
Gene Name
solute carrier family 16 member 6
Gene Aliases
MCT7
Chromosome Location
Chr.19: 61867803 - 61877805 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192672.3 NP_001179601.1 4 - 5 690 58

View Full Product Details