Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HCRTR2
Entrez Gene ID
529603
Gene Name
hypocretin receptor 2
Gene Aliases
-
Chromosome Location
Chr.23: 5119945 - 5251633 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192677.1 NP_001179606.1 5 - 6 990 100

View Full Product Details