Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CRIP3
Entrez Gene ID
530071
Gene Name
cysteine rich protein 3
Gene Aliases
-
Chromosome Location
Chr.23: 16894553 - 16902615 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192689.1 NP_001179618.1 6 - 7 494 74

View Full Product Details