Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KLHDC8A
Entrez Gene ID
532283
Gene Name
kelch domain containing 8A
Gene Aliases
-
Chromosome Location
Chr.16: 3100151 - 3105788 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192722.2 NP_001179651.1 3 - 4 760 86

View Full Product Details