Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MBNL3
Entrez Gene ID
540550
Gene Name
muscleblind like splicing regulator 3
Gene Aliases
-
Chromosome Location
Chr.X: 16204462 - 16325590 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192740.1 NP_001179669.1 7 - 8 962 99

View Full Product Details