Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
STC2
Entrez Gene ID
540573
Gene Name
stanniocalcin 2
Gene Aliases
-
Chromosome Location
Chr.20: 5000472 - 5011125 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192745.3 NP_001179674.2 2 - 3 301 72

View Full Product Details