Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PIP4K2A
Entrez Gene ID
533289
Gene Name
phosphatidylinositol-5-phosphate 4-kinase type 2 alpha
Gene Aliases
-
Chromosome Location
Chr.13: 23621605 - 23808162 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192769.1 NP_001179698.1 3 - 4 334 119

View Full Product Details