Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KCNC4
Entrez Gene ID
540768
Gene Name
potassium voltage-gated channel subfamily C member 4
Gene Aliases
-
Chromosome Location
Chr.3: 33194702 - 33218021 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192780.3 NP_001179709.1 1 - 2 756 77

View Full Product Details