Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LHX4
Entrez Gene ID
540797
Gene Name
LIM homeobox 4
Gene Aliases
LXH4
Chromosome Location
Chr.16: 61427758 - 61473954 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192785.3 NP_001179714.1 3 - 4 450 69
GenBank mRNA AB257754.1 - 1 - 2 94 69

View Full Product Details