Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CGRRF1
Entrez Gene ID
534344
Gene Name
cell growth regulator with ring finger domain 1
Gene Aliases
-
Chromosome Location
Chr.10: 67014641 - 67042099 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192807.1 NP_001179736.1 4 - 5 659 135

View Full Product Details