Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HTR5A
Entrez Gene ID
541200
Gene Name
5-hydroxytryptamine receptor 5A
Gene Aliases
-
Chromosome Location
Chr.4: 116984176 - 117001724 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192842.1 NP_001179771.1 1 - 2 744 73

View Full Product Details