Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
IDS
Entrez Gene ID
541272
Gene Name
iduronate 2-sulfatase
Gene Aliases
-
Chromosome Location
Chr.X: 31987330 - 32006650 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192851.1 NP_001179780.1 6 - 7 986 80

View Full Product Details