Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
MANEA
Entrez Gene ID
512853
Gene Name
mannosidase endo-alpha
Gene Aliases
-
Chromosome Location
Chr.9: 54296791 - 54379111 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001206725.1 NP_001193654.1 4 - 5 875 102
GenBank mRNA BC103138.1 - 4 - 5 877 102
JN385276.1 - 3 - 4 759 102
JQ946519.1 - 3 - 4 732 102
JQ946520.1 - 3 - 4 732 102
JQ946521.1 - 3 - 4 732 102
JQ946522.1 - 3 - 4 732 102

View Full Product Details