Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SNX30
Entrez Gene ID
517400
Gene Name
sorting nexin family member 30
Gene Aliases
-
Chromosome Location
Chr.8: 102004576 - 102108791 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001206738.1 NP_001193667.1 3 - 4 462 62

View Full Product Details