Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
OLIG1
Entrez Gene ID
539307
Gene Name
oligodendrocyte transcription factor 1
Gene Aliases
-
Chromosome Location
Chr.1: 2497798 - 2499871 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001206761.2 NP_001193690.1 1 - 1 1820 70

View Full Product Details