Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
UBXN7
Entrez Gene ID
539940
Gene Name
UBX domain protein 7
Gene Aliases
UBXD7
Chromosome Location
Chr.1: 70927372 - 70987463 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001206764.2 NP_001193693.1 4 - 5 385 91
GenBank mRNA EE353771.1 - 3 - 4 310 91

View Full Product Details