Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
IGSF5
Entrez Gene ID
511594
Gene Name
immunoglobulin superfamily member 5
Gene Aliases
-
Chromosome Location
Chr.1: 139827193 - 139881435 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001206778.1 NP_001193707.1 4 - 5 859 74

View Full Product Details