Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KRT14
Entrez Gene ID
404111
Gene Name
keratin 14
Gene Aliases
-
Chromosome Location
Chr.19: 41792379 - 41796743 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001166575.3 NP_001160047.3 1 - 2 717 88

View Full Product Details