Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GIP
Entrez Gene ID
511073
Gene Name
gastric inhibitory polypeptide
Gene Aliases
-
Chromosome Location
Chr.19: 37567749 - 37573010 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001166605.1 NP_001160077.1 5 - 6 481 65

View Full Product Details