Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KNOP1
Entrez Gene ID
511442
Gene Name
lysine rich nucleolar protein 1
Gene Aliases
C25H16orf88, TSG118
Chromosome Location
Chr.25: 17297693 - 17314276 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001166606.2 NP_001160078.1 3 - 4 1057 75
GenBank mRNA BC140578.1 - 2 - 3 982 75

View Full Product Details