Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RSPH10B
Entrez Gene ID
510922
Gene Name
radial spoke head 10 homolog B (Chlamydomonas)
Gene Aliases
-
Chromosome Location
Chr.25: 37980855 - 38006676 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001166614.1 NP_001160086.1 15 - 16 2035 115
GenBank mRNA BT025392.1 - 14 - 15 1984 115

View Full Product Details