Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RSPH10B
Entrez Gene ID
510922
Gene Name
radial spoke head 10 homolog B (Chlamydomonas)
Gene Aliases
-
Chromosome Location
Chr.25: 37980855 - 38006676 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001166614.1 NP_001160086.1 6 - 7 816 80

View Full Product Details