Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GORAB
Entrez Gene ID
614920
Gene Name
golgin, RAB6 interacting
Gene Aliases
NTKLBP1, SCYL1BP1
Chromosome Location
Chr.16: 38087696 - 38110825 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001167897.3 NP_001161369.1 1 - 2 141 94
GenBank mRNA BC142523.1 - 1 - 2 144 94

View Full Product Details