Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MRNIP
Entrez Gene ID
614757
Gene Name
MRN complex interacting protein
Gene Aliases
C7H5orf45
Chromosome Location
Chr.7: 1417994 - 1435526 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001167943.1 NP_001161415.1 5 - 6 500 86
GenBank mRNA BC103300.1 - 5 - 6 454 86
EH175421.1 - 5 - 6 500 86

View Full Product Details