Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KCNK5
Entrez Gene ID
782166
Gene Name
potassium two pore domain channel subfamily K member 5
Gene Aliases
-
Chromosome Location
Chr.23: 13060758 - 13101202 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191144.1 NP_001178073.1 4 - 5 948 82

View Full Product Details