Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PCDH10
Entrez Gene ID
531875
Gene Name
protocadherin 10
Gene Aliases
-
Chromosome Location
Chr.17: 25027051 - 25071224 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191151.1 NP_001178080.1 3 - 4 2862 74

View Full Product Details