Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
VWC2L
Entrez Gene ID
782360
Gene Name
von Willebrand factor C domain containing 2 like
Gene Aliases
-
Chromosome Location
Chr.2: 102410346 - 102617458 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191159.1 NP_001178088.1 2 - 3 525 81

View Full Product Details