Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ISY1
Entrez Gene ID
783414
Gene Name
ISY1 splicing factor homolog (S. cerevisiae)
Gene Aliases
-
Chromosome Location
Chr.22: 59047437 - 59063934 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191173.2 NP_001178102.1 7 - 8 553 59
GenBank mRNA BC142461.1 - 7 - 8 528 59

View Full Product Details