Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SAMD12
Entrez Gene ID
783254
Gene Name
sterile alpha motif domain containing 12
Gene Aliases
-
Chromosome Location
Chr.14: 45625075 - 46074096 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191180.2 NP_001178109.1 1 - 2 146 70

View Full Product Details