Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CTTNBP2NL
Entrez Gene ID
538981
Gene Name
CTTNBP2 N-terminal like
Gene Aliases
-
Chromosome Location
Chr.3: 30833767 - 30886450 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191181.2 NP_001178110.1 4 - 5 571 108
GenBank mRNA BC111189.1 - 7 - 8 736 108

View Full Product Details