Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
EOMES
Entrez Gene ID
511947
Gene Name
eomesodermin
Gene Aliases
-
Chromosome Location
Chr.22: 2109634 - 2115807 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191188.1 NP_001178117.1 4 - 5 1301 94

View Full Product Details