Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GTF2A1
Entrez Gene ID
785612
Gene Name
general transcription factor IIA subunit 1
Gene Aliases
-
Chromosome Location
Chr.10: 92473228 - 92511515 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191217.4 NP_001178146.2 1 - 2 217 106

View Full Product Details