Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ISL2
Entrez Gene ID
786913
Gene Name
ISL LIM homeobox 2
Gene Aliases
-
Chromosome Location
Chr.21: 31584548 - 31590222 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191237.2 NP_001178166.2 5 - 6 968 53

View Full Product Details