Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NLGN2
Entrez Gene ID
787986
Gene Name
neuroligin 2
Gene Aliases
-
Chromosome Location
Chr.19: 27083899 - 27098969 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191242.1 NP_001178171.1 3 - 4 661 85

View Full Product Details