Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ASIC3
Entrez Gene ID
789132
Gene Name
acid sensing ion channel subunit 3
Gene Aliases
ACCN3
Chromosome Location
Chr.4: 113624088 - 113629085 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191251.3 NP_001178180.3 4 - 5 1009 76

View Full Product Details