Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ZACN
Entrez Gene ID
789168
Gene Name
zinc activated ion channel
Gene Aliases
LGICZ1
Chromosome Location
Chr.19: 55578491 - 55582703 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191252.3 NP_001178181.1 6 - 7 669 57

View Full Product Details