Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PRDM14
Entrez Gene ID
100140032
Gene Name
PR/SET domain 14
Gene Aliases
-
Chromosome Location
Chr.14: 33786654 - 33805075 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191264.1 NP_001178193.1 1 - 2 704 69

View Full Product Details