Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MBD5
Entrez Gene ID
541084
Gene Name
methyl-CpG binding domain protein 5
Gene Aliases
-
Chromosome Location
Chr.2: 47696843 - 47853314 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191268.1 NP_001178197.1 9 - 10 4711 75

View Full Product Details