Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RBKS
Entrez Gene ID
513276
Gene Name
ribokinase
Gene Aliases
-
Chromosome Location
Chr.11: 71836069 - 71930877 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191271.1 NP_001178200.1 2 - 3 230 132

View Full Product Details