Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PLCL1
Entrez Gene ID
537873
Gene Name
phospholipase C like 1 (inactive)
Gene Aliases
-
Chromosome Location
Chr.2: 86319218 - 86688034 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191279.2 NP_001178208.1 2 - 3 3193 97

View Full Product Details