Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SGO2
Entrez Gene ID
532412
Gene Name
shugoshin 2
Gene Aliases
SGOL2
Chromosome Location
Chr.2: 89070468 - 89099327 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191290.3 NP_001178219.2 6 - 7 768 86
GenBank mRNA BC120219.1 - 6 - 7 772 86

View Full Product Details